Learn More
Meso Scale Discovery UPLEX Mouse IFN ASY384well
SDP
Catalog No. 5000081535
Encompass
The Meso Scale Discovery Human IFN-alpha Ultra-Sensitive Kit is an electrochemiluminescent immunoassay for measuring human interferon alpha (IFN-alpha) in various biological samples It uses MSD s SULFO-TAG labels and multi-array plate technology offering a detection limit of 0 04 pg/mL and a dynamic range up to 10 000 pg/mL The assay is specific for IFN-alpha with minimal cross-reactivity The kit includes a 96-well plate with capture antibody detection antibody calibrator and all necessary reagents and buffers
Catalog No. 50-000-81535 Supplier Meso Scale Discovery Supplier No. K252W1K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Human IFN-alpha Ultra-Sensitive Kit is an electrochemiluminescent immunoassay for measuring human interferon alpha (IFN-alpha) in various biological samples It uses MSD s SULFO-TAG labels and multi-array plate technology offering a detection limit of 0 04 pg/mL and a dynamic range up to 10 000 pg/mL The assay is specific for IFN-alpha with minimal cross-reactivity The kit includes a 96-well plate with capture antibody detection antibody calibrator and all necessary reagents and buffers