Learn More
Meso Scale Discovery UPLEX Mouse IFNB ASYQP 25 PL
SDP
Supplier:  Meso Scale Discovery K152G0K24
Encompass
The Meso Scale Discovery U-PLEX Mouse IFN beta Assay is a multiplex immunoassay kit for detecting and quantifying mouse interferon beta in various samples This electrochemiluminescent assay uses MSD s SECTOR Imager offering high sensitivity a broad dynamic range and low sample volume requirements The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls buffers diluent and seals It detects mouse IFN beta from 0 06 pg/mL to 2500 pg/mL
Catalog No. 50-000-80020
Add to Cart
Request a Quote