Learn More
Meso Scale Discovery UPLEX Mouse IL10 ASYQP 25 PL
SDP
Catalog No. 5000079333
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-10 Singleplex Assay is an electrochemiluminescence-based immunoassay designed for quantitative measurement of interleukin-10 in mouse samples This singleplex format allows focused detection of IL-10 a key anti-inflammatory cytokine using MSD s proprietary U-PLEX platform technology The assay provides researchers with a reliable tool for studying immune responses and inflammatory processes in mouse models with high sensitivity and specificity
Catalog No. 50-000-79333 Supplier Meso Scale Discovery Supplier No. K152TZK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-10 Singleplex Assay is an electrochemiluminescence-based immunoassay designed for quantitative measurement of interleukin-10 in mouse samples This singleplex format allows focused detection of IL-10 a key anti-inflammatory cytokine using MSD s proprietary U-PLEX platform technology The assay provides researchers with a reliable tool for studying immune responses and inflammatory processes in mouse models with high sensitivity and specificity