Learn More
Meso Scale Discovery UPLEX Mouse IL12/IL23p40 ASYQP
SDP
Catalog No. 5000078474
Encompass
The U-PLEX Mouse IL-12/IL-23p40 Antibody Set (B22UQ) is a specialized immunoassay kit from Mesoscale Discovery designed for quantitative detection of interleukin-12 and interleukin-23p40 cytokines in mouse samples This multiplex assay utilizes electrochemiluminescence technology to provide sensitive accurate measurements of these important inflammatory biomarkers in a single well format enabling efficient cytokine profiling for research applications
Catalog No. 50-000-78474 Supplier Meso Scale Discovery Supplier No. K152UQK21
The U-PLEX Mouse IL-12/IL-23p40 Antibody Set (B22UQ) is a specialized immunoassay kit from Mesoscale Discovery designed for quantitative detection of interleukin-12 and interleukin-23p40 cytokines in mouse samples This multiplex assay utilizes electrochemiluminescence technology to provide sensitive accurate measurements of these important inflammatory biomarkers in a single well format enabling efficient cytokine profiling for research applications