Learn More
Meso Scale Discovery UPLEX Mouse IL12p70 ASY384well
SDP
Catalog No. 5000081206
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-12p70 384-Well Assay Kit is a multiplex immunoassay for measuring mouse interleukin 12 (IL-12) in various biological samples It uses MSD s electrochemiluminescent detection technology and is compatible with the MESO QuickPlex SQ 120 instrument The kit includes a mouse IL-12p70-specific antibody on carbon electrodes in 384-well plates along with necessary calibrators and reagents Results are available in 3 5 hours with high sensitivity and a broad dynamic range offering a fast simple method to quantify IL-12p70 levels in mouse samples
Catalog No. 50-000-81206 Supplier Meso Scale Discovery Supplier No. K252UAK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-12p70 384-Well Assay Kit is a multiplex immunoassay for measuring mouse interleukin 12 (IL-12) in various biological samples It uses MSD s electrochemiluminescent detection technology and is compatible with the MESO QuickPlex SQ 120 instrument The kit includes a mouse IL-12p70-specific antibody on carbon electrodes in 384-well plates along with necessary calibrators and reagents Results are available in 3 5 hours with high sensitivity and a broad dynamic range offering a fast simple method to quantify IL-12p70 levels in mouse samples