Learn More
Meso Scale Discovery UPLEX Mouse IL13 ASYQP 25 PL
SDP
Catalog No. 5000078418
Encompass
Meso Scale Discovery offers IL-13 immunoassay products for research applications Their platform provides sensitive quantitative measurement of interleukin-13 a key cytokine involved in allergic responses and Th2 immune pathways The assays utilize electrochemiluminescence detection technology for enhanced sensitivity and dynamic range supporting both singleplex and multiplex formats for efficient biomarker analysis in various sample types
Catalog No. 50-000-78418 Supplier Meso Scale Discovery Supplier No. K152UBK24
Add to Cart
Add to Cart
Request a Quote
Meso Scale Discovery offers IL-13 immunoassay products for research applications Their platform provides sensitive quantitative measurement of interleukin-13 a key cytokine involved in allergic responses and Th2 immune pathways The assays utilize electrochemiluminescence detection technology for enhanced sensitivity and dynamic range supporting both singleplex and multiplex formats for efficient biomarker analysis in various sample types