Learn More
Meso Scale Discovery UPLEX Mouse IL17A ASY384well
SDP
Catalog No. 5000079439
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-17A 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 17A (IL-17A) in cell culture supernatants serum plasma and other biological fluids This kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay is provided in a 384-well plate format and includes a plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and controls The kit allows quantification of mouse IL-17A with a testing time of 3 5 hours and a sample volume as low as 25 L
Catalog No. 50-000-79439 Supplier Meso Scale Discovery Supplier No. K252UTK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-17A 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 17A (IL-17A) in cell culture supernatants serum plasma and other biological fluids This kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay is provided in a 384-well plate format and includes a plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and controls The kit allows quantification of mouse IL-17A with a testing time of 3 5 hours and a sample volume as low as 25 L