Learn More
Meso Scale Discovery UPLEX Mouse IL17C ASY384well
SDP
Catalog No. 5000081315
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-17C Assay Kit is a multiplex immunoassay that measures mouse interleukin 17C (IL-17C) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay kit includes a plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrator diluent wash buffer read buffer and a standard curve ranging from 3 2 pg/mL to 10 000 pg/mL The minimal detectable dose is 1 5 pg/mL This assay allows for the quantitative measurement of mouse IL-17C
Catalog No. 50-000-81315 Supplier Meso Scale Discovery Supplier No. K252WJK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-17C Assay Kit is a multiplex immunoassay that measures mouse interleukin 17C (IL-17C) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay kit includes a plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrator diluent wash buffer read buffer and a standard curve ranging from 3 2 pg/mL to 10 000 pg/mL The minimal detectable dose is 1 5 pg/mL This assay allows for the quantitative measurement of mouse IL-17C