Learn More
Meso Scale Discovery UPLEX Mouse IL17F ASY 25 PL
SDP
Catalog No. 5000082406
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-17F Assay Kit is a multiplex immunoassay for measuring mouse interleukin 17F (IL-17F) in various biological fluids It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The assay requires 50 L or less sample volume and is compatible with MSD instruments The kit includes a plate with capture antibodies detection antibodies and calibrator diluent It has a sensitivity of 0 3 pg/mL and an upper quantification limit of 10 000 pg/mL for natural and recombinant mouse IL-17F
Catalog No. 50-000-82406 Supplier Meso Scale Discovery Supplier No. K152WAK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-17F Assay Kit is a multiplex immunoassay for measuring mouse interleukin 17F (IL-17F) in various biological fluids It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The assay requires 50 L or less sample volume and is compatible with MSD instruments The kit includes a plate with capture antibodies detection antibodies and calibrator diluent It has a sensitivity of 0 3 pg/mL and an upper quantification limit of 10 000 pg/mL for natural and recombinant mouse IL-17F