Learn More
Meso Scale Discovery UPLEX Mouse IL17F ASY384well
SDP
Catalog No. 5000081536
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-17F 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 17F (IL-17F) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and assay diluent Results are obtained in as little as 3 5 hours using an MSD instrument like SECTOR Imager 2400 or QuickPlex SQ 120 The kit has high performance and is ideal for measuring IL-17F levels in mouse samples
Catalog No. 50-000-81536 Supplier Meso Scale Discovery Supplier No. K252WAK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-17F 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 17F (IL-17F) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and assay diluent Results are obtained in as little as 3 5 hours using an MSD instrument like SECTOR Imager 2400 or QuickPlex SQ 120 The kit has high performance and is ideal for measuring IL-17F levels in mouse samples