Learn More
Meso Scale Discovery UPLEX Mouse IL22 ASYQP 25 PL
SDP
Catalog No. 5000080332
Encompass
The U-PLEX Mouse IL-22 Assay (K152WIK) is a highly sensitive immunoassay kit designed to quantitatively measure interleukin-22 levels in mouse biological samples This electrochemiluminescence-based assay offers superior sensitivity and precision for cytokine detection in research applications The kit provides reliable reproducible results for studying IL-22 s role in immune responses inflammation and tissue homeostasis in mouse models making it essential for immunology and inflammatory disease research
Catalog No. 50-000-80332 Supplier Meso Scale Discovery Supplier No. K152WIK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Mouse IL-22 Assay (K152WIK) is a highly sensitive immunoassay kit designed to quantitatively measure interleukin-22 levels in mouse biological samples This electrochemiluminescence-based assay offers superior sensitivity and precision for cytokine detection in research applications The kit provides reliable reproducible results for studying IL-22 s role in immune responses inflammation and tissue homeostasis in mouse models making it essential for immunology and inflammatory disease research