Learn More
Meso Scale Discovery UPLEX Mouse IL27p28/IL30 ASY
SDP
Catalog No. 5000080421
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-27p28/IL-30 Assay Kit is a multiplex immunoassay that measures mouse IL-27p28 and IL-30 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on Meso Scale Discovery instrumentation including the MESO QuickPlex SQ 120 QuickPlex SQ 120 and V-PLEX Plus The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrator
Catalog No. 50-000-80421 Supplier Meso Scale Discovery Supplier No. K152WCK1
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Mouse IL-27p28/IL-30 Assay Kit is a multiplex immunoassay that measures mouse IL-27p28 and IL-30 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on Meso Scale Discovery instrumentation including the MESO QuickPlex SQ 120 QuickPlex SQ 120 and V-PLEX Plus The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrator