Learn More
Meso Scale Discovery UPLEX Mouse IL27p28/IL30 ASY
SDP
Catalog No. 5000082242
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-27p28/IL-30 Assay Kit is a multiplex immunoassay that measures mouse IL-27p28 and IL-30 in serum plasma and cell culture supernatants This assay uses MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG reagents calibrators and an MSD read buffer Sample incubation time is just 2 hours The kit can detect IL-27p28 in the range of 39 - 20 000 pg/mL and IL-30 in the range of 156 - 80 000 pg/mL
Catalog No. 50-000-82242 Supplier Meso Scale Discovery Supplier No. K152WCK2
The Meso Scale Discovery U-PLEX Mouse IL-27p28/IL-30 Assay Kit is a multiplex immunoassay that measures mouse IL-27p28 and IL-30 in serum plasma and cell culture supernatants This assay uses MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG reagents calibrators and an MSD read buffer Sample incubation time is just 2 hours The kit can detect IL-27p28 in the range of 39 - 20 000 pg/mL and IL-30 in the range of 156 - 80 000 pg/mL