Learn More
Meso Scale Discovery UPLEX Mouse IL31 ASYQP 25 PL
SDP
Catalog No. 5000081995
Encompass
The U-PLEX Mouse IL-31 Assay (K152WEK) is a highly sensitive electrochemiluminescence immunoassay designed to quantitatively measure interleukin-31 levels in mouse serum plasma and cell culture samples This multiplex-ready assay features a 10-fold lower sample volume requirement compared to traditional ELISA methods with a detection range of 0 61-2 500 pg/mL The kit includes pre-coated plates calibrators detection antibodies and read buffer for streamlined workflow Ideal for inflammation and immunology research applications
Catalog No. 50-000-81995 Supplier Meso Scale Discovery Supplier No. K152WEK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Mouse IL-31 Assay (K152WEK) is a highly sensitive electrochemiluminescence immunoassay designed to quantitatively measure interleukin-31 levels in mouse serum plasma and cell culture samples This multiplex-ready assay features a 10-fold lower sample volume requirement compared to traditional ELISA methods with a detection range of 0 61-2 500 pg/mL The kit includes pre-coated plates calibrators detection antibodies and read buffer for streamlined workflow Ideal for inflammation and immunology research applications