Learn More
Meso Scale Discovery UPLEX Mouse IL4 ASYQP 25 PL
SDP
Catalog No. 5000082150
Encompass
The MSD U-PLEX Mouse IL-4 Singleplex Assay is an electrochemiluminescence-based immunoassay kit designed for quantitative measurement of interleukin-4 protein levels in mouse biological samples This singleplex format allows focused analysis of IL-4 a key cytokine involved in immune responses and inflammation pathways The assay utilizes MSD s proprietary U-PLEX platform technology offering high sensitivity detection with minimal sample volume requirements and reduced assay time compared to traditional ELISA methods
Catalog No. 50-000-82150 Supplier Meso Scale Discovery Supplier No. K152TWK24
Add to Cart
Add to Cart
Request a Quote
The MSD U-PLEX Mouse IL-4 Singleplex Assay is an electrochemiluminescence-based immunoassay kit designed for quantitative measurement of interleukin-4 protein levels in mouse biological samples This singleplex format allows focused analysis of IL-4 a key cytokine involved in immune responses and inflammation pathways The assay utilizes MSD s proprietary U-PLEX platform technology offering high sensitivity detection with minimal sample volume requirements and reduced assay time compared to traditional ELISA methods