Learn More
Meso Scale Discovery UPLEX Mouse IL5 ASYQP 25 PL
SDP
Catalog No. 5000078667
Encompass
The U-PLEX Mouse IL-5 Assay (K152UOK-24) is a multiplex immunoassay kit from Meso Scale Discovery designed to quantitatively measure interleukin-5 levels in mouse biological samples This electrochemiluminescence-based assay provides high sensitivity and specificity for IL-5 detection supporting research in inflammation allergy and immune response studies The kit includes pre-coated plates detection antibodies and reagents for 24 tests offering researchers a reliable tool for cytokine analysis in preclinical mouse models
Catalog No. 50-000-78667 Supplier Meso Scale Discovery Supplier No. K152UOK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Mouse IL-5 Assay (K152UOK-24) is a multiplex immunoassay kit from Meso Scale Discovery designed to quantitatively measure interleukin-5 levels in mouse biological samples This electrochemiluminescence-based assay provides high sensitivity and specificity for IL-5 detection supporting research in inflammation allergy and immune response studies The kit includes pre-coated plates detection antibodies and reagents for 24 tests offering researchers a reliable tool for cytokine analysis in preclinical mouse models