Learn More
Meso Scale Discovery UPLEX Mouse Insulin ASY384well
SDP
Catalog No. 5000081217
Encompass
The Meso Scale Discovery U-PLEX Mouse Insulin 384-Well Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of insulin in mouse serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and buffer The assay has a sensitivity of 0 058 pg/mL and a range of 0 058-1500 pg/mL Sample volume is 25 L The total assay time is 3 5 hours This assay allows high-throughput quantification of insulin levels
Catalog No. 50-000-81217 Supplier Meso Scale Discovery Supplier No. K2526HK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse Insulin 384-Well Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of insulin in mouse serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and buffer The assay has a sensitivity of 0 058 pg/mL and a range of 0 058-1500 pg/mL Sample volume is 25 L The total assay time is 3 5 hours This assay allows high-throughput quantification of insulin levels