Learn More
Meso Scale Discovery UPLEX Mouse KC/GRO ASY384well
SDP
Catalog No. 5000081313
Encompass
The U-PLEX Mouse KC Assay Kit from Meso Scale Discovery is a multiplex immunoassay that enables simultaneous quantification of 10 mouse cytokines and chemokines (IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 IFN- TNF- and KC/GRO) in cell culture supernatants serum plasma and other biological matrices This electrochemiluminescent assay utilizes MSD s patented MULTI-ARRAY technology and is ideal for mouse inflammation studies and immunology research The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies read buffer calibrators and controls Sample incubation time is just 2 hours
Catalog No. 50-000-81313 Supplier Meso Scale Discovery Supplier No. K252VWK2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Mouse KC Assay Kit from Meso Scale Discovery is a multiplex immunoassay that enables simultaneous quantification of 10 mouse cytokines and chemokines (IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 IFN- TNF- and KC/GRO) in cell culture supernatants serum plasma and other biological matrices This electrochemiluminescent assay utilizes MSD s patented MULTI-ARRAY technology and is ideal for mouse inflammation studies and immunology research The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies read buffer calibrators and controls Sample incubation time is just 2 hours