Learn More
Meso Scale Discovery UPLEX Mouse Leptin ASY 25 PL
SDP
Catalog No. 5000079387
Encompass
The Meso Scale Discovery U-PLEX Mouse Leptin Assay Kit is a multiplex immunoassay for measuring mouse leptin in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity broad dynamic range and precision The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags calibrators and controls The assay takes about 3 5 hours and requires a Meso Scale Discovery instrument With a detection limit of 26 pg/mL it is a quantitative tool for studying leptin biology
Catalog No. 50-000-79387 Supplier Meso Scale Discovery Supplier No. K1525ZK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse Leptin Assay Kit is a multiplex immunoassay for measuring mouse leptin in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity broad dynamic range and precision The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags calibrators and controls The assay takes about 3 5 hours and requires a Meso Scale Discovery instrument With a detection limit of 26 pg/mL it is a quantitative tool for studying leptin biology