Learn More
Meso Scale Discovery UPLEX Mouse MCP1 ASYQP 25 PL
SDP
Catalog No. 5000078700
Encompass
MSD s U-PLEX Mouse MCP-1 assay is an electrochemiluminescence-based immunoassay for quantifying monocyte chemoattractant protein-1 in mouse samples This multiplex-compatible assay offers high sensitivity broad dynamic range and minimal sample volume requirements The U-PLEX platform enables researchers to measure multiple biomarkers simultaneously making it ideal for inflammation and immune response studies in mouse models
Catalog No. 50-000-78700 Supplier Meso Scale Discovery Supplier No. K152UGK24
Add to Cart
Add to Cart
Request a Quote
MSD s U-PLEX Mouse MCP-1 assay is an electrochemiluminescence-based immunoassay for quantifying monocyte chemoattractant protein-1 in mouse samples This multiplex-compatible assay offers high sensitivity broad dynamic range and minimal sample volume requirements The U-PLEX platform enables researchers to measure multiple biomarkers simultaneously making it ideal for inflammation and immune response studies in mouse models