Learn More
Meso Scale Discovery UPLEX Mouse MIP1 ASY384well
SDP
Catalog No. 5000081311
Encompass
The Meso Scale Discovery U-PLEX Mouse MIP-1alpha 384-well Assay Kit (K252UJK) is a multiplex immunoassay for measuring mouse MIP-1alpha in serum plasma and cell culture supernatants It uses electrochemiluminescence detection and provides reagents for 384 tests The assay has a sensitivity of 0 3 pg/mL and a 7-point standard curve from 0 3-10 000 pg/mL The kit includes a plate with capture antibodies ECL-tagged detection antibodies calibrators diluent read buffer wash buffer and adhesive seals enabling high-throughput sensitive quantification
Catalog No. 50-000-81311 Supplier Meso Scale Discovery Supplier No. K252UJK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse MIP-1alpha 384-well Assay Kit (K252UJK) is a multiplex immunoassay for measuring mouse MIP-1alpha in serum plasma and cell culture supernatants It uses electrochemiluminescence detection and provides reagents for 384 tests The assay has a sensitivity of 0 3 pg/mL and a 7-point standard curve from 0 3-10 000 pg/mL The kit includes a plate with capture antibodies ECL-tagged detection antibodies calibrators diluent read buffer wash buffer and adhesive seals enabling high-throughput sensitive quantification