Learn More
Meso Scale Discovery UPLEX Mouse MIP1B ASY 25 PL
SDP
Catalog No. 5000078473
Encompass
The Meso Scale Discovery U-PLEX Mouse MIP-1beta Assay is a multiplex immunoassay kit for quantifying mouse MIP-1beta in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity broad dynamic range and precision with minimal matrix interference The assay requires 50 L or less sample volume and is compatible with MESO QuickPlex SQ 120 SQ 120 Pro or SECTOR S 600 instruments The kit includes a plate with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and wash buffer allowing flexible analyte panel customization
Catalog No. 50-000-78473 Supplier Meso Scale Discovery Supplier No. K152UKK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse MIP-1beta Assay is a multiplex immunoassay kit for quantifying mouse MIP-1beta in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity broad dynamic range and precision with minimal matrix interference The assay requires 50 L or less sample volume and is compatible with MESO QuickPlex SQ 120 SQ 120 Pro or SECTOR S 600 instruments The kit includes a plate with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and wash buffer allowing flexible analyte panel customization