Learn More
Meso Scale Discovery UPLEX Mouse MIP1B ASY384well
SDP
Catalog No. 5000081259
Encompass
The Meso Scale Discovery U-PLEX Mouse MIP-1beta 384-Well Assay Kit (K252UKK) is a multiplex immunoassay that measures mouse MIP-1beta in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology in a 384-well plate format providing high sensitivity and a broad dynamic range The assay requires 10 L of sample per well and can be read on MSD instrument platforms The kit includes a plate coated with capture antibodies detection antibodies calibrators and diluent It allows for the quantitative determination of MIP-1beta concentrations
Catalog No. 50-000-81259 Supplier Meso Scale Discovery Supplier No. K252UKK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse MIP-1beta 384-Well Assay Kit (K252UKK) is a multiplex immunoassay that measures mouse MIP-1beta in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology in a 384-well plate format providing high sensitivity and a broad dynamic range The assay requires 10 L of sample per well and can be read on MSD instrument platforms The kit includes a plate coated with capture antibodies detection antibodies calibrators and diluent It allows for the quantitative determination of MIP-1beta concentrations