Learn More
Meso Scale Discovery UPLEX Mouse MIP3 ASY384well
SDP
Catalog No. 5000081207
Encompass
The Meso Scale Discovery U-PLEX Mouse MIP-3alpha 384-Well Assay Kit is a multiplex immunoassay for measuring mouse MIP-3alpha in serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample use The kit includes a 384-well plate with capture antibodies labeled detection antibodies calibrators and diluent It detects as low as 0 06 pg/mL and up to 10 000 pg/mL requiring 10 L per well with a total assay time of about 3 5 hours
Catalog No. 50-000-81207 Supplier Meso Scale Discovery Supplier No. K252UZK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse MIP-3alpha 384-Well Assay Kit is a multiplex immunoassay for measuring mouse MIP-3alpha in serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample use The kit includes a 384-well plate with capture antibodies labeled detection antibodies calibrators and diluent It detects as low as 0 06 pg/mL and up to 10 000 pg/mL requiring 10 L per well with a total assay time of about 3 5 hours