Learn More
Meso Scale Discovery UPLEX Mouse MMP9 total ASYQP
SDP
Catalog No. 5000082118
Encompass
The Meso Scale Discovery U-PLEX Mouse MMP-9 Total Assay Kit is a multiplex immunoassay for measuring MMP-9 in mouse samples like serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrator and diluent Sensitivity is 0 04 ng/mL with a quantification limit of 2500 ng/mL It requires 25 L per well and can assay 46 samples in duplicate in 3 5 hours
Catalog No. 50-000-82118 Supplier Meso Scale Discovery Supplier No. K152ZGK22
The Meso Scale Discovery U-PLEX Mouse MMP-9 Total Assay Kit is a multiplex immunoassay for measuring MMP-9 in mouse samples like serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrator and diluent Sensitivity is 0 04 ng/mL with a quantification limit of 2500 ng/mL It requires 25 L per well and can assay 46 samples in duplicate in 3 5 hours