Learn More
Meso Scale Discovery UPLEX Mouse RANTES ASY 25 PL
SDP
Catalog No. 5000082101
Encompass
The Meso Scale Discovery U-PLEX Mouse RANTES Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of RANTES in mouse biological samples such as serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance This assay allows rapid and reliable quantification of RANTES from mouse samples
Catalog No. 50-000-82101 Supplier Meso Scale Discovery Supplier No. K152A2K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse RANTES Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of RANTES in mouse biological samples such as serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance This assay allows rapid and reliable quantification of RANTES from mouse samples