Learn More
Meso Scale Discovery UPLEX Mouse RANTES ASY384well
SDP
Catalog No. 5000081195
Encompass
The Meso Scale Discovery U-PLEX Mouse RANTES 384-Well Assay K252A2K is a 384-well multiplex immunoassay kit for the quantitative measurement of RANTES in mouse samples such as serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a streamlined workflow This assay allows for the simultaneous measurement of mouse RANTES along with other biomarkers in the same sample to understand immune response With a testing volume of 25 L per well it is designed for high-throughput biomarker analysis studies
Catalog No. 50-000-81195 Supplier Meso Scale Discovery Supplier No. K252A2K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse RANTES 384-Well Assay K252A2K is a 384-well multiplex immunoassay kit for the quantitative measurement of RANTES in mouse samples such as serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a streamlined workflow This assay allows for the simultaneous measurement of mouse RANTES along with other biomarkers in the same sample to understand immune response With a testing volume of 25 L per well it is designed for high-throughput biomarker analysis studies