Learn More
Meso Scale Discovery UPLEX Mouse SDF1 ASY 25 PL
SDP
Catalog No. 5000078370
Encompass
The Meso Scale Discovery U-PLEX Mouse SDF-1alpha Assay is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of mouse SDF-1alpha in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and has a testing time of 3 5 hours The lower limit of detection is 1 5 pg/mL This product can aid in the investigation of SDF-1alpha biology and the effects of treatments on SDF-1alpha levels
Catalog No. 50-000-78370 Supplier Meso Scale Discovery Supplier No. K152VBK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse SDF-1alpha Assay is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of mouse SDF-1alpha in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and has a testing time of 3 5 hours The lower limit of detection is 1 5 pg/mL This product can aid in the investigation of SDF-1alpha biology and the effects of treatments on SDF-1alpha levels