Learn More
Meso Scale Discovery UPLEX Mouse SDF1 ASY384well
SDP
Catalog No. 5000081191
Encompass
The U-PLEX Mouse SDF-1alpha 384-well Assay Kit from Meso Scale Discovery is a 384-well immunoassay that quantitatively measures mouse SDF-1alpha (CXCL12) in serum plasma cell culture supernatants cell and tissue lysates This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and rapid time to results The kit includes a mouse SDF-1alpha standard and all necessary reagents and plates to perform the assay Results are analyzed on MSD s imaging systems like the MESO QuickPlex SQ 120 This assay allows for the accurate quantification of mouse SDF-1alpha across diverse biological samples
Catalog No. 50-000-81191 Supplier Meso Scale Discovery Supplier No. K252VBK2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Mouse SDF-1alpha 384-well Assay Kit from Meso Scale Discovery is a 384-well immunoassay that quantitatively measures mouse SDF-1alpha (CXCL12) in serum plasma cell culture supernatants cell and tissue lysates This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and rapid time to results The kit includes a mouse SDF-1alpha standard and all necessary reagents and plates to perform the assay Results are analyzed on MSD s imaging systems like the MESO QuickPlex SQ 120 This assay allows for the accurate quantification of mouse SDF-1alpha across diverse biological samples