Learn More
Meso Scale Discovery UPLEX Mouse TARC ASYQP 25 PL
SDP
Catalog No. 5000081375
Encompass
The U-PLEX Mouse TARC Assay from Meso Scale Discovery is a multiplex immunoassay that measures TARC (thymus and activation regulated chemokine) in mouse serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies and calibrators to generate a standard curve The assay takes 3 5 hours to complete and only requires 10 L of sample
Catalog No. 50-000-81375 Supplier Meso Scale Discovery Supplier No. K152B0K24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Mouse TARC Assay from Meso Scale Discovery is a multiplex immunoassay that measures TARC (thymus and activation regulated chemokine) in mouse serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies and calibrators to generate a standard curve The assay takes 3 5 hours to complete and only requires 10 L of sample