Learn More
Meso Scale Discovery UPLEX Mouse TGFB1 ASY 25 PL
SDP
Catalog No. 5000079112
Encompass
The Meso Scale Discovery U-PLEX Mouse TGF-beta1 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of TGF-beta1 in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent The assay takes approximately 3 5 hours to complete and requires 10 L of sample per well The lower limit of detection is 1 7 pg/mL
Catalog No. 50-000-79112 Supplier Meso Scale Discovery Supplier No. K152XWK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse TGF-beta1 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of TGF-beta1 in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent The assay takes approximately 3 5 hours to complete and requires 10 L of sample per well The lower limit of detection is 1 7 pg/mL