Learn More
Meso Scale Discovery UPLEX Mouse TGFB2 ASY384well
SDP
Catalog No. 5000081154
Encompass
The Meso Scale Discovery U-PLEX Mouse TGF-beta 2 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of TGF-beta 2 in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager to provide superior sensitivity and a broad dynamic range This kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized U-PLEX Calibrator The lower limit of detection is 1 3 pg/mL and the assay range is up to 10 000 pg/mL This product enables sensitive reproducible quantitation of mouse TGF-beta 2 for life science research applications
Catalog No. 50-000-81154 Supplier Meso Scale Discovery Supplier No. K252XUK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse TGF-beta 2 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of TGF-beta 2 in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager to provide superior sensitivity and a broad dynamic range This kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized U-PLEX Calibrator The lower limit of detection is 1 3 pg/mL and the assay range is up to 10 000 pg/mL This product enables sensitive reproducible quantitation of mouse TGF-beta 2 for life science research applications