Learn More
Meso Scale Discovery UPLEX Mouse TGFB3 ASY 25 PL
SDP
Catalog No. 5000082116
Encompass
The Meso Scale Discovery U-PLEX Mouse TGF-beta3 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of TGF-beta3 in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for measuring TGF-beta3 with a detection range of 1 5-1000 pg/mL and an assay time of 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent This assay allows for high-throughput screening and monitoring of TGF-beta3 levels in mouse models
Catalog No. 50-000-82116 Supplier Meso Scale Discovery Supplier No. K152XVK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse TGF-beta3 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of TGF-beta3 in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for measuring TGF-beta3 with a detection range of 1 5-1000 pg/mL and an assay time of 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent This assay allows for high-throughput screening and monitoring of TGF-beta3 levels in mouse models