Learn More
Meso Scale Discovery UPLEX Mouse TGFB3 ASY384well
SDP
Catalog No. 5000079977
Encompass
The Meso Scale Discovery U-PLEX Mouse TGF-beta3 384-Well Assay Kit is a multiplex immunoassay for measuring TGF-beta3 in mouse serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity broad dynamic range and precision The 384-well plate allows high throughput analysis of up to 80 samples with just 25 L The assay takes about 3 5 hours and requires an MSD instrument like SECTOR Imager or QuickPlex SQ 120 This sensitive reproducible kit is ideal for research on mouse TGF-beta3 levels
Catalog No. 50-000-79977 Supplier Meso Scale Discovery Supplier No. K252XVK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse TGF-beta3 384-Well Assay Kit is a multiplex immunoassay for measuring TGF-beta3 in mouse serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity broad dynamic range and precision The 384-well plate allows high throughput analysis of up to 80 samples with just 25 L The assay takes about 3 5 hours and requires an MSD instrument like SECTOR Imager or QuickPlex SQ 120 This sensitive reproducible kit is ideal for research on mouse TGF-beta3 levels