Learn More
Meso Scale Discovery UPLEX Mouse TNF ASY 25 PL
SDP
Catalog No. 5000079356
Encompass
The Meso Scale Discovery U-PLEX Mouse TNF-alpha Assay is a multiplex immunoassay kit for quantifying mouse TNF- in serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity broad dynamic range and precision with a 2 5-hour protocol Requiring only 25 L of sample it detects as low as 0 04 pg/mL The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators controls buffers and seals enabling simple and rapid quantification from various samples
Catalog No. 50-000-79356 Supplier Meso Scale Discovery Supplier No. K152UCK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse TNF-alpha Assay is a multiplex immunoassay kit for quantifying mouse TNF- in serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity broad dynamic range and precision with a 2 5-hour protocol Requiring only 25 L of sample it detects as low as 0 04 pg/mL The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators controls buffers and seals enabling simple and rapid quantification from various samples