Learn More
Meso Scale Discovery UPLEX Mouse TNF ASY384well
SDP
Catalog No. 5000081309
Encompass
The Meso Scale Discovery U-PLEX Mouse TNF-alpha 384-well Assay Kit is a multiplex immunoassay that measures mouse TNF-alpha in serum plasma cell culture supernatants cell and tissue lysates It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and buffer for 384 tests It can detect TNF-alpha concentrations as low as 0 04 pg/mL with an upper limit of 50 000 pg/mL
Catalog No. 50-000-81309 Supplier Meso Scale Discovery Supplier No. K252UCK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse TNF-alpha 384-well Assay Kit is a multiplex immunoassay that measures mouse TNF-alpha in serum plasma cell culture supernatants cell and tissue lysates It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and buffer for 384 tests It can detect TNF-alpha concentrations as low as 0 04 pg/mL with an upper limit of 50 000 pg/mL