Learn More
Meso Scale Discovery UPLEX Mouse TNFRI ASY 25 PL
SDP
Catalog No. 5000082144
Encompass
The Meso Scale Discovery U-PLEX Mouse TNF RI Assay Kit (K1520VK) is a multiplex immunoassay kit for the quantitative measurement of mouse tumor necrosis factor receptor 1 (TNFR1) in serum plasma and cell culture supernatants The assay uses electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantitation of 10 000 pg/mL for mouse TNFR1
Catalog No. 50-000-82144 Supplier Meso Scale Discovery Supplier No. K1520VK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse TNF RI Assay Kit (K1520VK) is a multiplex immunoassay kit for the quantitative measurement of mouse tumor necrosis factor receptor 1 (TNFR1) in serum plasma and cell culture supernatants The assay uses electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantitation of 10 000 pg/mL for mouse TNFR1