Learn More
Meso Scale Discovery UPLEX Mouse VEGFa ASY 25 PL
SDP
Catalog No. 5000078469
Encompass
The Meso Scale Discovery U-PLEX Mouse VEGF-A Assay Kit is a multiplex immunoassay that measures mouse VEGF-A in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can measure VEGF-A concentrations as low as 0 06 pg/mL with as little as 25 L of sample It includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and an MSD read buffer The total assay time is 3 5 hours
Catalog No. 50-000-78469 Supplier Meso Scale Discovery Supplier No. K152UVK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse VEGF-A Assay Kit is a multiplex immunoassay that measures mouse VEGF-A in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can measure VEGF-A concentrations as low as 0 06 pg/mL with as little as 25 L of sample It includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and an MSD read buffer The total assay time is 3 5 hours