Learn More
Meso Scale Discovery UPLEX Mouse VEGFa ASY384well
SDP
Catalog No. 5000081312
Encompass
The Meso Scale Discovery U-PLEX Mouse VEGF-A 384-Well Assay Kit is a multiplex immunoassay that measures mouse VEGF-A in serum plasma cell culture supernatants and other biological fluids It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra-/inter-assay precision This 384-well plate assay allows users to quantify mouse VEGF-A levels with as little as 25 L of sample The kit includes a VEGF-A standard detection antibody Read Buffer and 384-well plate coated with capture antibodies It can detect mouse VEGF-A with a lower limit of detection of 0 06 pg/mL and an upper limit of 10 000 pg/mL
Catalog No. 50-000-81312 Supplier Meso Scale Discovery Supplier No. K252UVK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse VEGF-A 384-Well Assay Kit is a multiplex immunoassay that measures mouse VEGF-A in serum plasma cell culture supernatants and other biological fluids It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra-/inter-assay precision This 384-well plate assay allows users to quantify mouse VEGF-A levels with as little as 25 L of sample The kit includes a VEGF-A standard detection antibody Read Buffer and 384-well plate coated with capture antibodies It can detect mouse VEGF-A with a lower limit of detection of 0 06 pg/mL and an upper limit of 10 000 pg/mL