Learn More
Meso Scale Discovery UPLEX MS Combo 1HMN
SDP
Catalog No. 5000080842
Encompass
The Meso Scale Discovery U-PLEX Multiple Sclerosis Combo 1 Human Sector K15684K is a multiplex assay kit that allows simultaneous measurement of IL-2 IL-10 IL-17A IL-17F IFN- and TNF- in human serum plasma and cell culture supernatants This electrochemiluminescence (ECL) based assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent calibrators and controls This assay allows efficient quantification of multiple analytes from a single small-volume sample
Catalog No. 50-000-80842 Supplier Meso Scale Discovery Supplier No. K15684K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Multiple Sclerosis Combo 1 Human Sector K15684K is a multiplex assay kit that allows simultaneous measurement of IL-2 IL-10 IL-17A IL-17F IFN- and TNF- in human serum plasma and cell culture supernatants This electrochemiluminescence (ECL) based assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent calibrators and controls This assay allows efficient quantification of multiple analytes from a single small-volume sample