Learn More
Meso Scale Discovery UPLEX MS Combo 1HMN
SDP
Catalog No. 5000081059
Encompass
The Meso Scale Discovery U-PLEX Multiple Sclerosis Combo 1 Human Sector Kit K15684K is a multiplex immunoassay kit for the simultaneous detection of human cytokines and chemokines The kit contains a 10-spot MULTI-ARRAY plate pre-coated with capture antibodies for 10 analytes IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 and TNF- The kit includes detection antibodies and calibrators to generate standard curves and quantify analyte concentrations in serum plasma and cell culture supernatants This kit allows sensitive reproducible measurement of multiple analytes related to multiple sclerosis from a single small sample volume
Catalog No. 50-000-81059 Supplier Meso Scale Discovery Supplier No. K15684K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Multiple Sclerosis Combo 1 Human Sector Kit K15684K is a multiplex immunoassay kit for the simultaneous detection of human cytokines and chemokines The kit contains a 10-spot MULTI-ARRAY plate pre-coated with capture antibodies for 10 analytes IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 and TNF- The kit includes detection antibodies and calibrators to generate standard curves and quantify analyte concentrations in serum plasma and cell culture supernatants This kit allows sensitive reproducible measurement of multiple analytes related to multiple sclerosis from a single small sample volume