Learn More
Meso Scale Discovery UPLEX MSE 6CKine/CCL21 ASY
SDP
Catalog No. 5000082211
Encompass
The Meso Scale Discovery U-PLEX Mouse 6CKine (CCL21) Assay is a multiplex immunoassay kit that enables simultaneous quantification of 6 mouse cytokines and chemokines (IL-2 IL-4 IL-5 IL-10 IFN- and CCL21) from cell culture supernatants serum plasma and other biological samples It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay requires an MSD instrument such as SECTOR Imager for plate reading It can help monitor immune responses and is suitable for mouse samples
Catalog No. 50-000-82211 Supplier Meso Scale Discovery Supplier No. K152C0K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse 6CKine (CCL21) Assay is a multiplex immunoassay kit that enables simultaneous quantification of 6 mouse cytokines and chemokines (IL-2 IL-4 IL-5 IL-10 IFN- and CCL21) from cell culture supernatants serum plasma and other biological samples It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay requires an MSD instrument such as SECTOR Imager for plate reading It can help monitor immune responses and is suitable for mouse samples