Learn More
Meso Scale Discovery UPLEX MSE 6CKine/CCL21 ASYQP
SDP
Catalog No. 5000081355
Encompass
The Meso Scale Discovery U-PLEX Mouse 6CKine (CCL21) Assay is a multiplex immunoassay that measures mouse CCL21 and 5 other cytokines in cell culture supernatants serum plasma and other biological samples It uses electrochemiluminescence detection technology for high sensitivity and broad dynamic range The assay requires 50 L or less of sample and can be read on Meso Scale Discovery instrumentation including the MESO QuickPlex SQ 120 The assay measures CCL21 GM-CSF IFN- IL-10 IL-12p70 and TNF- simultaneously It has high specificity and low cross-reactivity The assay takes 3 5 hours to complete
Catalog No. 50-000-81355 Supplier Meso Scale Discovery Supplier No. K152C0K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse 6CKine (CCL21) Assay is a multiplex immunoassay that measures mouse CCL21 and 5 other cytokines in cell culture supernatants serum plasma and other biological samples It uses electrochemiluminescence detection technology for high sensitivity and broad dynamic range The assay requires 50 L or less of sample and can be read on Meso Scale Discovery instrumentation including the MESO QuickPlex SQ 120 The assay measures CCL21 GM-CSF IFN- IL-10 IL-12p70 and TNF- simultaneously It has high specificity and low cross-reactivity The assay takes 3 5 hours to complete