Learn More
Meso Scale Discovery UPLEX MSE 6CKine/CCL2ASY384
SDP
Catalog No. 5000079445
Encompass
The Meso Scale Discovery U-PLEX Mouse 6CKine (CCL21) 384-well Assay Kit is designed to measure CCL21 in mouse samples such as serum plasma and cell culture supernatants This multiplex immunoassay kit allows simultaneous quantification of CCL21 and 5 other cytokines or chemokines The kit includes a 384-well plate coated with capture antibodies detection antibodies and calibrators The electrochemiluminescent detection method provides high sensitivity wide dynamic range and low sample volume requirements This assay kit can help researchers studying chemokine biology and immunology in mouse models
Catalog No. 50-000-79445 Supplier Meso Scale Discovery Supplier No. K252C0K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse 6CKine (CCL21) 384-well Assay Kit is designed to measure CCL21 in mouse samples such as serum plasma and cell culture supernatants This multiplex immunoassay kit allows simultaneous quantification of CCL21 and 5 other cytokines or chemokines The kit includes a 384-well plate coated with capture antibodies detection antibodies and calibrators The electrochemiluminescent detection method provides high sensitivity wide dynamic range and low sample volume requirements This assay kit can help researchers studying chemokine biology and immunology in mouse models