Learn More
Meso Scale Discovery UPLEX MSE 6CKine/CCL2ASY384
SDP
Catalog No. 5000081211
Encompass
The Meso Scale Discovery U-PLEX Mouse 6CKine (CCL21) 384-well Assay Kit is designed to measure CCL21 in mouse samples It is a multiplex immunoassay that uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This kit includes a 384-well plate coated with capture antibodies for mouse CCL21 detection antibodies labeled with an ECL tag calibrators and diluent The assay takes approximately 3 5 hours to complete and each kit contains enough reagents for 1 x 384-well plate This quantitative immunoassay allows simultaneous detection of mouse CCL21 with a lower limit of detection of 17 2 pg/mL
Catalog No. 50-000-81211 Supplier Meso Scale Discovery Supplier No. K252C0K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse 6CKine (CCL21) 384-well Assay Kit is designed to measure CCL21 in mouse samples It is a multiplex immunoassay that uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This kit includes a 384-well plate coated with capture antibodies for mouse CCL21 detection antibodies labeled with an ECL tag calibrators and diluent The assay takes approximately 3 5 hours to complete and each kit contains enough reagents for 1 x 384-well plate This quantitative immunoassay allows simultaneous detection of mouse CCL21 with a lower limit of detection of 17 2 pg/mL