Learn More
Meso Scale Discovery UPLEX MSE BAFF ASY384well
SDP
Catalog No. 5000081252
Encompass
The Meso Scale Discovery Mouse BAFF U-PLEX Assay is a multiplex immunoassay panel that measures BAFF (B cell activating factor) in mouse biological samples such as serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume This assay allows for the simultaneous measurement of mouse BAFF along with other biomarkers It can be used in mouse studies related to B cell biology immunology and autoimmunity The assay requires the Meso Scale Discovery SECTOR Imager and provides results in approximately 3 hours
Catalog No. 50-000-81252 Supplier Meso Scale Discovery Supplier No. K2523QK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Mouse BAFF U-PLEX Assay is a multiplex immunoassay panel that measures BAFF (B cell activating factor) in mouse biological samples such as serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume This assay allows for the simultaneous measurement of mouse BAFF along with other biomarkers It can be used in mouse studies related to B cell biology immunology and autoimmunity The assay requires the Meso Scale Discovery SECTOR Imager and provides results in approximately 3 hours