Learn More
Meso Scale Discovery UPLEX MSE BCA1/BLC ASY384well
SDP
Catalog No. 5000079326
Encompass
The Meso Scale Discovery Mouse BCA-1/BLC Singleplex Assay Kit is a singleplex immunoassay that allows for the quantitative measurement of BCA-1/BLC in mouse samples such as serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The assay can detect BCA-1/BLC concentrations as low as 3 2 pg/mL with an upper limit of 10 000 pg/mL The total assay time is less than 3 5 hours for 96-well plates This product can aid in the quantitative determination of BCA-1/BLC levels in mouse biological samples
Catalog No. 50-000-79326 Supplier Meso Scale Discovery Supplier No. K252F0K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Mouse BCA-1/BLC Singleplex Assay Kit is a singleplex immunoassay that allows for the quantitative measurement of BCA-1/BLC in mouse samples such as serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The assay can detect BCA-1/BLC concentrations as low as 3 2 pg/mL with an upper limit of 10 000 pg/mL The total assay time is less than 3 5 hours for 96-well plates This product can aid in the quantitative determination of BCA-1/BLC levels in mouse biological samples