Learn More
Meso Scale Discovery UPLEX MSE BCA1/BLC ASY384well
SDP
Catalog No. 5000079444
Encompass
The Meso Scale Discovery BCA-1 BLC Kit is a highly sensitive electrochemiluminescent immunoassay for the quantitative determination of B-cell activating factor (BAFF) in human serum plasma and cell culture supernatants The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide superior sensitivity a wide dynamic range and high-throughput capabilities for BAFF detection The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to ruthenium label calibrators and assay diluent The assay has a typical sensitivity of 1 5 pg/mL and an upper limit of quantification of 1000 pg/mL
Catalog No. 50-000-79444 Supplier Meso Scale Discovery Supplier No. K252F0K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery BCA-1 BLC Kit is a highly sensitive electrochemiluminescent immunoassay for the quantitative determination of B-cell activating factor (BAFF) in human serum plasma and cell culture supernatants The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide superior sensitivity a wide dynamic range and high-throughput capabilities for BAFF detection The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to ruthenium label calibrators and assay diluent The assay has a typical sensitivity of 1 5 pg/mL and an upper limit of quantification of 1000 pg/mL