Learn More
Meso Scale Discovery UPLEX MSE BCA1/BLC ASYQP
SDP
Catalog No. 5000081448
Encompass
The Meso Scale Discovery U-PLEX Mouse BCA-1/BLC Assay is a multiplex immunoassay kit for measuring BCA-1 and BLC in mouse samples like serum plasma and cell culture supernatants Using MSD s SULFO-TAG labels and multi-array technology it offers sensitive detection from a small sample in 3 5 hours The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators controls buffers plate seals and an MSD instrument It ensures a streamlined workflow and rapid results for BCA-1 and BLC quantification
Catalog No. 50-000-81448 Supplier Meso Scale Discovery Supplier No. K152F0K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse BCA-1/BLC Assay is a multiplex immunoassay kit for measuring BCA-1 and BLC in mouse samples like serum plasma and cell culture supernatants Using MSD s SULFO-TAG labels and multi-array technology it offers sensitive detection from a small sample in 3 5 hours The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators controls buffers plate seals and an MSD instrument It ensures a streamlined workflow and rapid results for BCA-1 and BLC quantification