Learn More
Meso Scale Discovery UPLEX MSE BDNF ASY384well
SDP
Catalog No. 5000079443
Encompass
The Meso Scale Discovery Mouse BDNF Kit is a 4-plate kit for the quantitative measurement of BDNF in mouse samples It uses electrochemiluminescence (ECL) technology and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer sample diluent calibrator diluent wash buffer and a vial of lyophilized calibrator The kit can accurately quantify BDNF across a range of 15 6-2000 pg/mL in mouse serum plasma tissue lysates and cell culture supernatants
Catalog No. 50-000-79443 Supplier Meso Scale Discovery Supplier No. K2526WK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Mouse BDNF Kit is a 4-plate kit for the quantitative measurement of BDNF in mouse samples It uses electrochemiluminescence (ECL) technology and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer sample diluent calibrator diluent wash buffer and a vial of lyophilized calibrator The kit can accurately quantify BDNF across a range of 15 6-2000 pg/mL in mouse serum plasma tissue lysates and cell culture supernatants